ENA018104752 Enamine

4-methoxy-3-{[(pyridin-4-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD03655063
  • Cas Number : 750601-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00667
Availability details for
Quantity
  • Availability
  • Add to Cart