ENA018104774 Enamine

2-(4-Chloro-phenyl)-quinoline-4-carboxylic acid

  • MDL number MFCD00085413
  • Cas Number : 5466-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00689

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00689
Availability details for
Quantity
  • Availability
  • Add to Cart