ENA018104790 Enamine

2-{5-[(2-carboxyphenyl)sulfamoyl]-2,4-dimethoxybenzenesulfonamido}benzoic acid

  • MDL number MFCD06621946
  • Cas Number : 825600-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00706
Availability details for
Quantity
  • Availability
  • Add to Cart