ENA018104799 Enamine

5,7-dibromo-1-benzofuran-2-carboxylic acid

  • MDL number MFCD00094315
  • Cas Number : 90415-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 275-276
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00718
Availability details for
Quantity
  • Availability
  • Add to Cart