ENA018104874 Enamine

4-hydroxy-2,6-diphenyl-4H-1,4lambda5-oxaphosphinin-4-one

  • MDL number MFCD01017116
  • Cas Number : 56153-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00806
Availability details for
Quantity
  • Availability
  • Add to Cart