ENA018104878 Enamine

1-(4-fluorophenyl)-3-methyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD02711496
  • Cas Number : 325721-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-210
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00810
Availability details for
Quantity
  • Availability
  • Add to Cart