ENA018104881 Enamine

2-(3,4-dimethoxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD00219544
  • Cas Number : 133676-16-1
  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-216
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00813
Availability details for
Quantity
  • Availability
  • Add to Cart