ENA018104888 Enamine

3-(3-methoxyphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02725466
  • Cas Number : 381214-37-5
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-methoxyphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C17H14N2O3/c1-22-14-9-5-6-12(10-14)16-15(17(20)21)11-19(18-16)13-7-3-2-4-8-13/h2-11H,1H3,(H,20,21)
Inchi Key YUDUFBFUQOJLGG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-00820

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-00820
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart