ENA018104903 Enamine

3-Benzylidene-2,3-dihydro-1H-cyclopenta[b]quinoline-9-carboxylic acid

  • MDL number MFCD03152477
  • Cas Number : 22311-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00841
Availability details for
Quantity
  • Availability
  • Add to Cart