ENA018104921 Enamine

4-Chloro-1,3-diphenyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD02714151
  • Cas Number : 377766-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00859
Availability details for
Quantity
  • Availability
  • Add to Cart