ENA018104922 Enamine

(2-Oxo-5-trifluoromethyl-2H-pyridin-1-yl)-acetic acid

  • MDL number MFCD02073538
  • Cas Number : 308294-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00862
Availability details for
Quantity
  • Availability
  • Add to Cart