ENA018104930 Enamine

2-oxo-6-thia-1,8-diazatricyclo[7.4.0.0,3,7]trideca-3(7),4,8,10,12-pentaene-5-carboxylic acid

  • MDL number MFCD02725386
  • Cas Number : 380348-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00870
Availability details for
Quantity
  • Availability
  • Add to Cart