ENA018104939 Enamine

2-Chloro-quinoline-4-carbonyl chloride

  • MDL number MFCD06654888
  • Cas Number : 2388-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00880
Availability details for
Quantity
  • Availability
  • Add to Cart