ENA018104950 Enamine

3-(4-Diethylamino-benzylidene)-2,3-dihydro-1H-cyclopenta[b]quinoline-9-carboxylic acid

  • MDL number MFCD03964202
  • Cas Number : 726157-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00891
Availability details for
Quantity
  • Availability
  • Add to Cart