ENA018104961 Enamine

2-tert-Butyl-4-(3-chloro-benzylidene)-1,2,3,4-tetrahydro-acridine-9-carboxylic acid

  • MDL number MFCD03476309
  • Cas Number : 380436-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00902
Availability details for
Quantity
  • Availability
  • Add to Cart