ENA018104962 Enamine

7-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD03478360
  • Cas Number : 438031-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00903
Availability details for
Quantity
  • Availability
  • Add to Cart