ENA018104965 Enamine

4-(3-Bromo-benzylidene)-2-methyl-1,2,3,4-tetrahydro-benzo[b][1,6]naphthyridine-10-carboxylic acid

  • MDL number MFCD03479900
  • Cas Number : 519151-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00906
Availability details for
Quantity
  • Availability
  • Add to Cart