ENA018104973 Enamine

2-ethyl-1H,2H,3H,4H-benzo[b]1,6-naphthyridine-10-carboxylic acid

  • MDL number MFCD03479190
  • Cas Number : 519150-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00914
Availability details for
Quantity
  • Availability
  • Add to Cart