ENA018104979 Enamine

3-(4-Chloro-benzylidene)-2,3-dihydro-1H-cyclopenta[b]quinoline-9-carboxylic acid

  • MDL number MFCD03651682
  • Cas Number : 522624-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00920
Availability details for
Quantity
  • Availability
  • Add to Cart