ENA018104981 Enamine

3,5-Dimethyl-1H-pyrrole-2,4-dicarboxylic acid 4-ethyl ester

  • MDL number MFCD00030408
  • Cas Number : 5442-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00922

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-00922
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart