ENA018104987 Enamine

3,7-Dichloro-6-methoxy-benzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD00449853
  • Cas Number : 34576-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...109
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00929

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00929
Availability details for
Quantity
  • Availability
  • Add to Cart