ENA018105002 Enamine

2,5-dimethyl-1-(3-methylphenyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD03658004
  • Cas Number : 749219-01-0
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00945
Availability details for
Quantity
  • Availability
  • Add to Cart