ENA018105027 Enamine

1-(2,5-Dichloro-phenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02629507
  • Cas Number : 568543-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00970

Safety Information

msds https://www.enaminestore.com/catalog/EN300-00970
Availability details for
Quantity
  • Availability
  • Add to Cart