ENA018105034 Enamine

6-methylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD02253172
  • Cas Number : 75001-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00977
Availability details for
Quantity
  • Availability
  • Add to Cart