ENA018105035 Enamine

3,6-dimethylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD02707983
  • Cas Number : 130182-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00978
Availability details for
Quantity
  • Availability
  • Add to Cart