ENA018105036 Enamine

6-phenylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD00139592
  • Cas Number : 74630-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00979
Availability details for
Quantity
  • Availability
  • Add to Cart