ENA018105041 Enamine

2-methyl-3-[2-(pyridin-4-yl)ethyl]indolizine-1-carbaldehyde

  • MDL number MFCD02707613
  • Cas Number : 327085-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00985
Availability details for
Quantity
  • Availability
  • Add to Cart