ENA018105048 Enamine

2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)acetaldehyde

  • Cas Number : 84-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00993
s_phrases_eu 26-36
Availability details for
Quantity
  • Availability
  • Add to Cart