ENA018105050 Enamine

4-(2,4-dimethoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02049275
  • Cas Number : 23111-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-00996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00996
Availability details for
Quantity
  • Availability
  • Add to Cart