ENA018105076 Enamine

1-benzyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00187663
  • Cas Number : 1217-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01023
Availability details for
Quantity
  • Availability
  • Add to Cart