ENA018105090 Enamine

4-ethoxy-2-hydroxybenzaldehyde

  • MDL number MFCD03032734
  • Cas Number : 43057-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-35
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01037
Availability details for
Quantity
  • Availability
  • Add to Cart