ENA018105091 Enamine

2-Hydroxy-4-propoxy-benzaldehyde

  • MDL number MFCD03147298
  • Cas Number : 63667-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01038
Availability details for
Quantity
  • Availability
  • Add to Cart