ENA018105100 Enamine

3-(3,4-Dimethoxy-phenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02918326
  • Cas Number : 381230-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01048

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01048
Availability details for
Quantity
  • Availability
  • Add to Cart