ENA018105105 Enamine

1-(propan-2-yl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00224231
  • Cas Number : 10487-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-01054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01054
Availability details for
Quantity
  • Availability
  • Add to Cart