ENA018105107 Enamine

methyl 4-[(2-formylphenoxy)methyl]benzoate

  • MDL number MFCD01893190
  • Cas Number : 351335-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01056
Availability details for
Quantity
  • Availability
  • Add to Cart