ENA018105110 Enamine

1-Pentyl-1H-indole-2,3-dione

  • MDL number MFCD00224228
  • Cas Number : 4290-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01059
Availability details for
Quantity
  • Availability
  • Add to Cart