ENA018105112 Enamine

1-(4-ethoxyphenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02213020
  • Cas Number : 347331-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01061
Availability details for
Quantity
  • Availability
  • Add to Cart