ENA018105118 Enamine

1-{[4-(2-hydroxyethyl)piperazin-1-yl]methyl}-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00411970
  • Cas Number : 6532-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-113
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01069
Availability details for
Quantity
  • Availability
  • Add to Cart