ENA018105130 Enamine

3-methyl-5-phenoxy-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02711484
  • Cas Number : 30241-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01085
Availability details for
Quantity
  • Availability
  • Add to Cart