ENA018105133 Enamine

2,4-bis(difluoromethoxy)benzaldehyde

  • MDL number MFCD03147271
  • Cas Number : 80395-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01088
Availability details for
Quantity
  • Availability
  • Add to Cart