ENA018105143 Enamine

N-(4-Acetyl-phenyl)-4-chloro-benzenesulfonamide

  • MDL number MFCD00424664
  • Cas Number : 72178-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01098
Availability details for
Quantity
  • Availability
  • Add to Cart