ENA018105149 Enamine

4-(3-phenyl-4,5-dihydro-1H-pyrazol-1-yl)benzaldehyde

  • MDL number MFCD00491118
  • Cas Number : 961-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01104
Availability details for
Quantity
  • Availability
  • Add to Cart