ENA018105169 Enamine

5-(4-fluorophenoxy)-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03950729
  • Cas Number : 926257-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01124
Availability details for
Quantity
  • Availability
  • Add to Cart