ENA018105171 Enamine

2-(1,3-benzothiazol-2-ylsulfanyl)-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD02323221
  • Cas Number : 405924-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01126
Availability details for
Quantity
  • Availability
  • Add to Cart