ENA018105172 Enamine

4-oxo-2-[(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD02673364
  • Cas Number : 746610-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-01127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01127
Availability details for
Quantity
  • Availability
  • Add to Cart