ENA018105174 Enamine

7-Bromo-5-fluoro-1H-indole-2,3-dione

  • MDL number MFCD03150727
  • Cas Number : 380431-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215...217
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01129

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-01129
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart