ENA018105199 Enamine

1-(2-ethoxyethyl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00469859
  • Cas Number : 303065-45-4
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01154
Availability details for
Quantity
  • Availability
  • Add to Cart