ENA018105216 Enamine

4-Difluoromethoxy-3-methoxy-benzaldehyde

  • MDL number MFCD03412217
  • Cas Number : 162401-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01171

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-01171
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart