ENA018105265 Enamine

3-Isothiocyanato-N,N-dimethyl-benzenesulfonamide

  • MDL number MFCD03147330
  • Cas Number : 379255-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01236
Availability details for
Quantity
  • Availability
  • Add to Cart