ENA018105270 Enamine

(2-isothiocyanatoethyl)dimethylamine

  • MDL number MFCD03147279
  • Cas Number : 7097-89-4
  • Molecular Weight : 130.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-01252

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P280,P285,P301+P312,P304+P341,P305+P351+P338,P310,P330,P342+P311,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-01252
pictograms GHS05,GHS07,GHS08
hstatements H227,H302,H318,H334
Availability details for
Quantity
  • Availability
  • Add to Cart